Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHGDH
Entrez Gene ID
100144529
Gene Name
phosphoglycerate dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.4: 101445977 - 101478144 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123162.1 NP_001116634.1 11 - 12 1596 60
GenBank mRNA AK239145.1 - 11 - 12 1548 60
AK240308.1 - 8 - 9 1180 60
AK392790.1 - 11 - 12 1546 60
AY609518.1 - 11 - 12 1527 60

Gene Symbol
LOC100156167
Entrez Gene ID
100156167
Gene Name
D-3-phosphoglycerate dehydrogenase-like
Gene Aliases
-
Chromosome Location
Chr.4: 101408108 - 101415914 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001926763.4 XP_001926798.4 - 744 60

View Full Product Details