Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LEP
Entrez Gene ID
396832
Gene Name
leptin
Gene Aliases
OB, OBS
Chromosome Location
Chr.18: 20106867 - 20124071 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213840.1 NP_999005.1 1 - 2 975 68
XM_021078500.1 XP_020934159.1 - 234 68
XM_021078502.1 XP_020934161.1 - 285 68
GenBank mRNA AF026976.2 - 2 - 3 203 68
AF052691.1 - 1 - 2 975 68
AF102856.1 - 1 - 2 149 68
GQ240885.1 - 1 - 2 149 68
GQ268935.1 - 1 - 2 158 68
GQ268936.1 - 1 - 2 150 68
HM194603.1 - 1 - 2 149 68
HM194604.1 - 1 - 2 149 68
U40812.1 - 1 - 2 107 68
U59894.1 - 1 - 2 149 68
U63540.1 - 1 - 2 182 68

View Full Product Details