Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CS
Entrez Gene ID
397519
Gene Name
citrate synthase
Gene Aliases
-
Chromosome Location
Chr.5: 21669778 - 21700737 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214276.1 NP_999441.1 8 - 9 924 68
XM_021091143.1 XP_020946802.1 - 1053 68
XM_021091144.1 XP_020946803.1 - 1209 68
XM_021091145.1 XP_020946804.1 - 1194 68
XM_021091147.1 XP_020946806.1 - 737 68
GenBank mRNA AK231644.1 - 8 - 9 1106 68
AK235732.1 - 9 - 10 1138 68
AK236734.1 - 9 - 10 1137 68
AK238572.1 - 9 - 10 1136 68
AK389252.1 - 7 - 8 804 68
AK389361.1 - 8 - 9 1101 68
AK390176.1 - 8 - 9 1147 68
AK394047.1 - 9 - 10 1139 68
AK395560.1 - 9 - 10 1156 68
AK396408.1 - 8 - 9 1044 68
AK399051.1 - - 1449 68
AK400607.1 - 8 - 9 1066 68
AK400978.1 - 8 - 9 1061 68
DQ403143.1 - 2 - 3 252 68
M21197.1 - 8 - 9 924 68

View Full Product Details