Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CS
Entrez Gene ID
397519
Gene Name
citrate synthase
Gene Aliases
-
Chromosome Location
Chr.5: 21669778 - 21700737 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214276.1 NP_999441.1 9 - 10 1026 92
XM_021091143.1 XP_020946802.1 - 1155 92
XM_021091144.1 XP_020946803.1 - 1311 92
XM_021091145.1 XP_020946804.1 - 1296 92
XM_021091147.1 XP_020946806.1 - 839 92
GenBank mRNA AK231644.1 - 9 - 10 1208 92
AK235732.1 - 10 - 11 1240 92
AK236734.1 - 10 - 11 1239 92
AK238572.1 - 10 - 11 1238 92
AK389252.1 - 8 - 9 906 92
AK389361.1 - 9 - 10 1203 92
AK390176.1 - 9 - 10 1249 92
AK394047.1 - 10 - 11 1241 92
AK395560.1 - 10 - 11 1258 92
AK396408.1 - 9 - 10 1146 92
AK399051.1 - - 1551 92
AK400607.1 - 9 - 10 1168 92
AK400978.1 - 9 - 10 1163 92
DQ403143.1 - 3 - 4 354 92
M21197.1 - 9 - 10 1026 92

View Full Product Details