Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSME2
Entrez Gene ID
397522
Gene Name
proteasome activator subunit 2
Gene Aliases
-
Chromosome Location
Chr.7: 75154570 - 75157871 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214279.1 NP_999444.1 2 - 3 141 96
GenBank mRNA AF527991.1 - 2 - 3 141 96
AK232253.1 - 2 - 3 144 96
AK234031.1 - 2 - 3 136 96
AK346956.1 - 2 - 3 144 96

View Full Product Details