Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP5F1E
Entrez Gene ID
100134963
Gene Name
ATP synthase F1 subunit epsilon
Gene Aliases
ATP5E
Chromosome Location
Chr.17: 59163085 - 59174488 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113448.1 NP_001106919.1 2 - 2 1851 112

View Full Product Details