Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANGPTL7
Entrez Gene ID
100217376
Gene Name
angiopoietin like 7
Gene Aliases
-
Chromosome Location
Chr.6: 71350872 - 71356299 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142828.2 NP_001136300.1 5 - 5 1053 73
GenBank mRNA FJ432080.1 - 5 - 5 1056 73

View Full Product Details