Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLVCR1
Entrez Gene ID
100217398
Gene Name
FLVCR heme transporter 1
Gene Aliases
-
Chromosome Location
Chr.9: 130543911 - 130570486 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142846.1 NP_001136318.1 1 - 1 351 72
GenBank mRNA FJ436992.1 - 1 - 1 351 72

View Full Product Details