Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMNDC1
Entrez Gene ID
100127466
Gene Name
survival motor neuron domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 120726077 - 120739673 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166309.1 NP_001159781.1 1 - 2 198 87
XM_005671451.3 XP_005671508.1 - 186 87
GenBank mRNA AK232698.1 - 1 - 2 174 87
AY609548.1 - 1 - 2 151 87
EU571478.1 - 1 - 2 198 87

View Full Product Details