Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL28B
Entrez Gene ID
100310828
Gene Name
interleukin 28B (interferon, lambda 3)
Gene Aliases
IL-28b
Chromosome Location
Chr.6: 47951925 - 47953282 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166490.1 NP_001159962.1 1 - 2 187 67
GenBank mRNA GQ996936.1 - 1 - 2 187 67

Gene Symbol
LOC110255217
Entrez Gene ID
110255217
Gene Name
interferon lambda-3-like
Gene Aliases
-
Chromosome Location
Chr.6: 47939706 - 47941254 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021097830.1 XP_020953489.1 - 187 67

View Full Product Details