Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FSCN1
Entrez Gene ID
100286741
Gene Name
fascin actin-bundling protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 4134173 - 4143275 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146300.2 NP_001139772.1 3 - 4 1234 90
GenBank mRNA AK236699.1 - 3 - 4 1236 90
AK392031.1 - 3 - 4 1232 90
FJ790418.1 - 3 - 4 1236 90

View Full Product Details