Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ARF3
Entrez Gene ID
100301570
Gene Name
ADP ribosylation factor 3
Gene Aliases
-
Chromosome Location
Chr.5: 14973477 - 14993016 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160426.1 NP_001153898.1 2 - 3 259 75
XM_021090975.1 XP_020946634.1 - 604 75
GenBank mRNA AK346946.1 - 3 - 4 513 75
AK390432.1 - 3 - 4 531 75

View Full Product Details