Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDHA
Entrez Gene ID
407245
Gene Name
lactate dehydrogenase A
Gene Aliases
LDH-A, LDH-M
Chromosome Location
Chr.2: 40814163 - 40827284 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172363.2 NP_001165834.1 4 - 5 532 94
XM_013994501.2 XP_013849955.2 - 1182 94
XM_021081651.1 XP_020937310.1 - 615 94
XM_021081655.1 XP_020937314.1 - 923 94
XM_021081659.1 XP_020937318.1 - 744 94
GenBank mRNA AK236433.1 - 4 - 5 570 94
AK343612.1 - 4 - 5 523 94
AK343811.1 - 4 - 5 521 94
AK391205.1 - 4 - 5 523 94
AK391208.1 - 4 - 5 520 94
AK391657.1 - 4 - 5 524 94
AK394718.1 - 2 - 3 289 94
AK395255.1 - 4 - 5 556 94
AK399432.1 - 4 - 5 524 94
AK399447.1 - 4 - 5 556 94
AK399472.1 - 4 - 5 539 94
AK399544.1 - 4 - 5 523 94
AK399555.1 - 4 - 5 523 94
AK399560.1 - 6 - 7 732 94
FJ865398.1 - 4 - 5 533 94
U07178.1 - 4 - 5 519 94

View Full Product Details