Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOGA1
Entrez Gene ID
100153961
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
DHDPSL
Chromosome Location
Chr.14: 108958812 - 108986373 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190169.1 NP_001177098.1 5 - 6 1058 83
GenBank mRNA AK236562.1 - 5 - 6 1058 83
AK393618.1 - 5 - 6 726 83

View Full Product Details