Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H4orf47
Entrez Gene ID
100154862
Gene Name
chromosome 15 C4orf47 homolog
Gene Aliases
C4orf47
Chromosome Location
Chr.15: 46470680 - 46612173 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190210.1 NP_001177139.1 7 - 8 1105 81
XM_005671711.2 XP_005671768.1 - 1238 81
XM_005671712.2 XP_005671769.1 - 1171 81
XM_005671713.3 XP_005671770.1 - 1077 81
XM_005671714.3 XP_005671771.1 - 1892 81
XM_013984044.2 XP_013839498.1 - 1201 81
XM_013984045.2 XP_013839499.1 - 1071 81
XM_021074525.1 XP_020930184.1 - 1196 81
GenBank mRNA AK239144.1 - 7 - 8 1102 81

View Full Product Details