Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REG4
Entrez Gene ID
100154580
Gene Name
regenerating family member 4
Gene Aliases
-
Chromosome Location
Chr.4: 101337295 - 101354550 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190251.1 NP_001177180.1 1 - 2 42 95
GenBank mRNA AY610013.1 - 1 - 2 60 95
EV992326.2 - 1 - 2 56 95

View Full Product Details