Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK20
Entrez Gene ID
100157041
Gene Name
cyclin dependent kinase 20
Gene Aliases
-
Chromosome Location
Chr.14: 15582 - 24102 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195329.1 NP_001182258.1 1 - 2 79 65
XM_005670379.3 XP_005670436.2 - 408 65
XM_013982617.2 XP_013838071.2 - 408 65
GenBank mRNA AK347973.1 - 1 - 2 331 65
GU373708.1 - 1 - 2 79 65

Gene Symbol
LOC100513119
Entrez Gene ID
100513119
Gene Name
cyclin-dependent kinase 20-like
Gene Aliases
-
Chromosome Location
Chr.Unknown: 18383 - 25797 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013984684.2 XP_013840138.2 - 400 65

View Full Product Details