Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGALS7
Entrez Gene ID
100217394
Gene Name
galectin 7
Gene Aliases
LGALS7B
Chromosome Location
Chr.6: 47588983 - 47604520 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142843.1 NP_001136315.1 2 - 3 112 65
XM_021093249.1 XP_020948908.1 - 863 65
XM_021093250.1 XP_020948909.1 - 1272 65
GenBank mRNA FJ424705.1 - 2 - 3 112 65

View Full Product Details