Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBCB
Entrez Gene ID
100522808
Gene Name
tubulin folding cofactor B
Gene Aliases
CKAP1
Chromosome Location
Chr.6: 45482542 - 45492627 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205403.1 NP_001192332.1 5 - 6 701 66
GenBank mRNA AK240439.1 - 5 - 6 693 66
AK352168.1 - 5 - 6 674 66
AK397066.1 - - 1067 66
CJ016107.1 - 5 - 6 701 66
EV946445.2 - 2 - 3 97 66

View Full Product Details