Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCO2
Entrez Gene ID
100516804
Gene Name
SCO cytochrome oxidase deficient homolog 2 (yeast)
Gene Aliases
-
Chromosome Location
Chr.5: 220801 - 222678 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206376.1 NP_001193305.1 2 - 3 346 119

View Full Product Details