Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YKT6
Entrez Gene ID
100513964
Gene Name
YKT6 v-SNARE homolog
Gene Aliases
-
Chromosome Location
Chr.18: 50961451 - 50971384 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244914.1 NP_001231843.1 5 - 6 947 61
GenBank mRNA AK233852.1 - 5 - 6 947 61
AY609946.1 - 5 - 6 509 61

View Full Product Details