Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNT16
Entrez Gene ID
100513992
Gene Name
polypeptide N-acetylgalactosaminyltransferase 16
Gene Aliases
GALNTL1
Chromosome Location
Chr.7: 92928834 - 93041446 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244918.1 NP_001231847.1 10 - 11 1220 109
XM_005666324.3 XP_005666381.1 - 1369 109
GenBank mRNA AK349556.1 - 10 - 11 1220 109

View Full Product Details