Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTRF1L
Entrez Gene ID
100514202
Gene Name
mitochondrial translational release factor 1 like
Gene Aliases
-
Chromosome Location
Chr.1: 13443602 - 13458291 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244934.1 NP_001231863.1 1 - 2 281 97
XM_013992473.2 XP_013847927.2 - 3186 97
GenBank mRNA AY609949.1 - 1 - 2 281 97

View Full Product Details