Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIH1D1
Entrez Gene ID
100514581
Gene Name
PIH1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 54524172 - 54529700 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244978.1 NP_001231907.1 8 - 9 1147 83
XM_005664715.3 XP_005664772.1 - 975 83
XM_005664716.3 XP_005664773.1 - 972 83
GenBank mRNA AK234303.1 - 8 - 9 1147 83
AK236765.1 - 8 - 9 1165 83
AK348445.1 - 8 - 9 1147 83
AY609770.1 - 12 - 13 1396 83

View Full Product Details