Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GHDC
Entrez Gene ID
100523395
Gene Name
GH3 domain containing
Gene Aliases
-
Chromosome Location
Chr.12: 20580937 - 20588662 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246239.1 NP_001233168.1 8 - 9 1518 89
XM_005668808.3 XP_005668865.1 - 1728 89
XM_005668812.3 XP_005668869.1 - 1548 89
GenBank mRNA HM241658.1 - 8 - 9 1457 89
HM241659.1 - 7 - 8 1639 89
HM241660.1 - 8 - 9 1518 89
HM241661.1 - 7 - 8 1178 89
HM241662.1 - 7 - 8 1264 89

View Full Product Details