Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNPNAT1
Entrez Gene ID
100152942
Gene Name
glucosamine-phosphate N-acetyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.1: 182404460 - 182423453 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243376.1 NP_001230305.1 3 - 4 425 110
XM_005659948.3 XP_005660005.1 - 369 110
XM_013993391.2 XP_013848845.1 - 470 110
GenBank mRNA AK343636.1 - 3 - 4 321 110
AY609449.1 - 3 - 4 279 110

View Full Product Details