Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACTB2
Entrez Gene ID
100153212
Gene Name
lactamase beta 2
Gene Aliases
-
Chromosome Location
Chr.4: 64710777 - 64748951 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243410.1 NP_001230339.1 1 - 2 186 101
GenBank mRNA AK344685.1 - 1 - 2 157 101
AK389783.1 - 1 - 2 144 101
AK392309.1 - 1 - 2 187 101
FS668818.1 - 1 - 2 187 101

View Full Product Details