Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R8
Entrez Gene ID
100627843
Gene Name
protein phosphatase 1 regulatory subunit 8
Gene Aliases
-
Chromosome Location
Chr.6: 84966746 - 84990664 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243414.1 NP_001230343.1 4 - 5 553 87
XM_021093434.1 XP_020949093.1 - 298 87
GenBank mRNA AK239489.1 - 4 - 5 509 87
BP464075.1 - 4 - 5 513 87
FD600058.1 - 5 - 6 553 87

View Full Product Details