Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMIGD3
Entrez Gene ID
104564308
Gene Name
transmembrane and immunoglobulin domain containing 3
Gene Aliases
-
Chromosome Location
Chr.4: 108814405 - 108871101 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243436.2 NP_001230365.1 7 - 8 860 72
GenBank mRNA CX056678.1 - 4 - 5 355 72

View Full Product Details