Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA3
Entrez Gene ID
100154010
Gene Name
pleckstrin homology domain containing A3
Gene Aliases
-
Chromosome Location
Chr.15: 84185007 - 84206887 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243464.1 NP_001230393.1 7 - 8 791 86
GenBank mRNA AK344147.1 - 4 - 5 441 86
EW635931.2 - 4 - 5 440 86

View Full Product Details