Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC15
Entrez Gene ID
100626939
Gene Name
thioredoxin domain containing 15
Gene Aliases
-
Chromosome Location
Chr.2: 137060988 - 137079288 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243618.1 NP_001230547.1 4 - 5 919 82
GenBank mRNA AK239456.1 - 4 - 5 919 82
AK345466.1 - 2 - 3 309 82
AK350186.1 - 5 - 6 852 82
AK396939.1 - 4 - 5 1253 82

View Full Product Details