Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCEA1
Entrez Gene ID
100157724
Gene Name
transcription elongation factor A1
Gene Aliases
-
Chromosome Location
Chr.4: 77109555 - 77138324 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243680.1 NP_001230609.1 5 - 6 589 81
XM_005653062.3 XP_005653119.1 - 657 81
GenBank mRNA AK349980.1 - 5 - 6 589 81

View Full Product Details