Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPQ
Entrez Gene ID
100157979
Gene Name
carboxypeptidase Q
Gene Aliases
PGCP
Chromosome Location
Chr.4: 39492663 - 39855959 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243691.1 NP_001230620.1 5 - 6 1065 79
GenBank mRNA AK349998.1 - 5 - 6 1065 79

View Full Product Details