Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MON1A
Entrez Gene ID
100626383
Gene Name
MON1 homolog A, secretory trafficking associated
Gene Aliases
-
Chromosome Location
Chr.13: 32426733 - 32437735 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243701.1 NP_001230630.1 5 - 6 1674 95
GenBank mRNA AK346293.1 - 5 - 6 1674 95
AK390410.1 - 5 - 6 1801 95

View Full Product Details