Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN3
Entrez Gene ID
100158159
Gene Name
MORN repeat containing 3
Gene Aliases
-
Chromosome Location
Chr.14: 30991897 - 31006578 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243709.1 NP_001230638.1 6 - 7 960 76
GenBank mRNA AK351364.1 - 6 - 7 960 76

View Full Product Details