Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARM1
Entrez Gene ID
100511208
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 70788134 - 70892642 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243716.1 NP_001230645.1 1 - 2 312 61
GenBank mRNA AK231710.1 - 1 - 2 314 61
AK350405.1 - 1 - 2 252 61

View Full Product Details