Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KLHDC4
Entrez Gene ID
100511452
Gene Name
kelch domain containing 4
Gene Aliases
-
Chromosome Location
Chr.6: 1627054 - 1672328 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243840.1 NP_001230769.1 10 - 11 1579 64
GenBank mRNA AK240179.1 - 10 - 11 1579 64

View Full Product Details