Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCARB2
Entrez Gene ID
414407
Gene Name
scavenger receptor class B member 2
Gene Aliases
-
Chromosome Location
Chr.8: 71838395 - 71910461 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244155.1 NP_001231084.1 8 - 9 1432 88
XM_005666776.2 XP_005666833.1 - 1427 88
GenBank mRNA AY609583.1 - 8 - 9 1232 88

View Full Product Details