Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SCARB2
- Entrez Gene ID
-
414407
- Gene Name
- scavenger receptor class B member 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 71838395 - 71910461 on Build Sscrofa11.1
- Gene Symbol
- AASDH
- Entrez Gene ID
-
100627475
- Gene Name
- aminoadipate-semialdehyde dehydrogenase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 55604394 - 55645536 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK391323.1 |
- |
- |
3250 |
87 |
View Full Product Details