Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRLF3
Entrez Gene ID
100525951
Gene Name
cytokine receptor like factor 3
Gene Aliases
-
Chromosome Location
Chr.12: 42933417 - 42991395 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244329.1 NP_001231258.1 4 - 5 670 78
XM_005669062.3 XP_005669119.2 - 695 78
XM_021066135.1 XP_020921794.1 - 698 78
GenBank mRNA AK343440.1 - 4 - 5 670 78
AK351417.1 - 4 - 5 601 78
CJ019492.1 - 1 - 2 118 78
DB787462.1 - 4 - 5 670 78

View Full Product Details