Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCP1A
Entrez Gene ID
100623102
Gene Name
decapping mRNA 1A
Gene Aliases
-
Chromosome Location
Chr.13: 35392592 - 35455639 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244358.1 NP_001231287.1 1 - 2 140 109
GenBank mRNA AK230977.1 - 1 - 2 244 109
AK237144.1 - 1 - 2 159 109
AK238754.1 - 1 - 2 159 109
AK347683.1 - 1 - 2 140 109
AK399200.1 - 1 - 2 174 109

View Full Product Details