Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCN2
Entrez Gene ID
100152846
Gene Name
transcobalamin 2
Gene Aliases
-
Chromosome Location
Chr.14: 47496245 - 47515966 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244436.1 NP_001231365.1 1 - 2 162 77
XM_005670817.3 XP_005670874.1 - 153 77
XM_005670820.3 XP_005670877.1 - 153 77
XM_005670821.3 XP_005670878.1 - 153 77
XM_013983049.2 XP_013838503.1 - 154 77
XM_021071885.1 XP_020927544.1 - 153 77
GenBank mRNA AK234715.1 - 1 - 2 105 77

View Full Product Details