Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRM1
Entrez Gene ID
100513493
Gene Name
mitochondrial rRNA methyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.12: 38123505 - 38143894 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244822.1 NP_001231751.1 3 - 4 979 99
XM_021066102.1 XP_020921761.1 - 1875 99
XM_021066103.1 XP_020921762.1 - 1875 99
GenBank mRNA AK345151.1 - 3 - 4 979 99
AK392643.1 - 2 - 3 1139 99

View Full Product Details