Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOR
Entrez Gene ID
100232842
Gene Name
glutathione reductase (NADPH)
Gene Aliases
GSVIVT01000729001
Chromosome Location
Chr.7: 475110 - 482501 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF019907.1 - 7 - 8 1345 85

View Full Product Details