Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MDH
Entrez Gene ID
100232859
Gene Name
malate dehydrogenase
Gene Aliases
GSVIVT00035904001
Chromosome Location
Chr.10: 1235479 - 1240575 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF195869.1 - 5 - 6 875 72

View Full Product Details