Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MADS4
Entrez Gene ID
100232869
Gene Name
MADS-box protein 4
Gene Aliases
GSVIVT00002777001
Chromosome Location
Chr.1_NW_002238160: 86453 - 103957 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF373603.1 - 2 - 3 357 87

View Full Product Details