Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MADS5
Entrez Gene ID
100232870
Gene Name
MADS-box protein 5
Gene Aliases
-
Chromosome Location
Chr.18: 18299365 - 18307232 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF373604.1 - 7 - 8 579 79

View Full Product Details