Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIP32
Entrez Gene ID
100232888
Gene Name
ripening-related protein-like
Gene Aliases
-
Chromosome Location
Chr.8: 2532500 - 2533616 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ237991.1 - 1 - 2 435 136

View Full Product Details