Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XT1
Entrez Gene ID
100232898
Gene Name
Beta-1,2-xylosyltransferase
Gene Aliases
GSVIVT00027221001
Chromosome Location
Chr.19: 7582324 - 7586998 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AM179856.1 - 2 - 3 691 77

View Full Product Details