Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BR6OX1
Entrez Gene ID
100232983
Gene Name
brassinosteroid-6-oxidase
Gene Aliases
GSVIVT01036558001
Chromosome Location
Chr.14: 12633743 - 12636889 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ235273.1 - 2 - 3 552 65

View Full Product Details